Anti-AGR2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NBP1-05933B
NOVUNBP1-05933BEA 687 EUR
NOVUNBP1-05933B
Anti-AGR2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
The AG-2 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to AG-2. This antibody reacts with human, mouse. The AG-2 Antibody [Biotin] has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: AG-2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR