Anti-VPRBP Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-05953
NOVUNBP1-05953EA 684 EUR
NOVUNBP1-05953
Anti-VPRBP Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal VprBP Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: VPRBP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human