Anti-KDM6B Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Novus Biologicals
NBP1-06640C
NOVUNBP1-06640CEA 768 EUR
NOVUNBP1-06640C
Anti-KDM6B Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: KDM6B (lysine (K)-specific demethylase 6B)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR