Anti-TNF-alpha Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-19532
NOVUNBP1-19532EA 732 EUR
NOVUNBP1-19532
Anti-TNF-alpha Rabbit Polyclonal Antibody
Antikörper
The TNF-alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to TNF-alpha. This antibody reacts with human, mouse, rat. The TNF-alpha Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50 to 1:200

Type: Primary
Antigen: TNF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR