Anti-B3GAT1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-19788B
NOVUNBP1-19788BEA 668 EUR
NOVUNBP1-19788B
Anti-B3GAT1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat