Anti-B3GAT1 Rabbit Polyclonal Antibody (DyLight® 488)
Lieferant: Novus Biologicals
NBP1-19788G
NOVUNBP1-19788GEA 668 EUR
NOVUNBP1-19788G
Anti-B3GAT1 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat