Anti-SCD Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Novus Biologicals
NBP1-26393F
NOVUNBP1-26393FEA 668 EUR
NOVUNBP1-26393F
Anti-SCD Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: SCD (stearoyl-CoA desaturase (delta-9-desaturase))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human