Anti-Del-1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-28632B
NOVUNBP1-28632BEA 722 EUR
NOVUNBP1-28632B
Anti-Del-1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Del-1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse