Anti-TYRO3 Rabbit Polyclonal Antibody (DyLight® 650)
Lieferant: Novus Biologicals
NBP1-28635C
NOVUNBP1-28635CEA 722 EUR
NOVUNBP1-28635C
Anti-TYRO3 Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: TYRO3 (TYRO3 protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse