Anti-PROX1 mouse monoclonal antibody
Lieferant: Novus Biologicals
NBP1-30045
NOVUNBP1-30045EA 638 EUR
NOVUNBP1-30045
Anti-PROX1 mouse monoclonal antibody
Antikörper
The Prox1 Antibody (5G10) from Novus Biologicals is a mouse monoclonal antibody to Prox1. This antibody reacts with human, mouse, rat, chicken, feline (negative). The Prox1 Antibody (5G10) has been validated for the following applications: Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount, CyTOF-ready.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000
Type: Primary
Antigen: Prox1
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope: The homeoProspero domain of Prox1
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken, Feline (Negative)