Anti-HIF-PH3 Mouse Monoclonal Antibody [clone: EG188e/d5] (DyLight® 550)

Lieferant: Novus Biologicals
NBP1-30440R
NOVUNBP1-30440REA 715 EUR
NOVUNBP1-30440R
Anti-HIF-PH3 Mouse Monoclonal Antibody [clone: EG188e/d5] (DyLight® 550)
Antikörper
Mouse Monoclonal HIF Prolyl Hydroxylase 3 Antibody (EG188e/d5) [DyLight 550]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Primate.

Type: Primary
Antigen: HIF-PH3
Clonality: Monoclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR