Anti-CD74 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-33109
NOVUNBP1-33109EA 702 EUR
NOVUNBP1-33109
Anti-CD74 Rabbit Polyclonal Antibody
Antikörper
The CD74 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CD74. This antibody reacts with human, mouse, rat. The CD74 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 to 1:500
Type: Primary
Antigen: CD74
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat