Anti-GAD2 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-33284
NOVUNBP1-33284EA 740 EUR
NOVUNBP1-33284
Anti-GAD2 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal GAD65 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: GAD65
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR