Anti-HIF1A Rabbit Polyclonal Antibody (Biotin) [clone: exon 9]
Lieferant: Novus Biologicals
NBP1-47178B
NOVUNBP1-47178BEA 668 EUR
NOVUNBP1-47178B
Anti-HIF1A Rabbit Polyclonal Antibody (Biotin) [clone: exon 9]
Antikörper
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone: exon 9
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human