Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 550) [clone: exon 13]

Lieferant: Novus Biologicals
NBP1-47181R
NOVUNBP1-47181REA 524 EUR
NOVUNBP1-47181R
Anti-HIF1A Rabbit Polyclonal Antibody (DyLight® 550) [clone: exon 13]
Antikörper
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone: exon 13
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR