Anti-ESRRA Rabbit Polyclonal Antibody (DyLight® 550)

Lieferant: Novus Biologicals
NBP1-47254R
NOVUNBP1-47254REA 668 EUR
NOVUNBP1-47254R
Anti-ESRRA Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Primary
Antigen: ESRRA (estrogen-related receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR