Anti-EphB4 mouse monoclonal antibody [clone: 5B5]

Lieferant: Novus Biologicals
NBP1-47434
NOVUNBP1-47434EA 588 EUR
NOVUNBP1-47434
Anti-EphB4 mouse monoclonal antibody [clone: 5B5]
Antikörper
The EphB4 Antibody (5B5) from Novus Biologicals is a mouse monoclonal antibody to EphB4. This antibody reacts with human. The EphB4 Antibody (5B5) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: EphB4
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR