Anti-PKM Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-48308B
NOVUNBP1-48308BEA 722 EUR
NOVUNBP1-48308B
Anti-PKM Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat