Anti-PKM Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Novus Biologicals
NBP1-48308F
NOVUNBP1-48308FEA 722 EUR
NOVUNBP1-48308F
Anti-PKM Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: PKM (pyruvate kinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat