Anti-N Cadherin Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 13A9]

Lieferant: Novus Biologicals
NBP1-48309F
NOVUNBP1-48309FEA 722 EUR
NOVUNBP1-48309F
Anti-N Cadherin Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 13A9]
Antikörper
Type: Primary
Antigen: N Cadherin
Clonality: monoclonal
Clone: 13A9
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR