Anti-C12orf5 Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Novus Biologicals
NBP1-49534G
NOVUNBP1-49534GEA 668 EUR
NOVUNBP1-49534G
Anti-C12orf5 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: C12orf5 (chromosome 12 open reading frame 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR