Anti-RBBP6 Rabbit Polyclonal Antibody (DyLight® 405)

Lieferant: Novus Biologicals
NBP1-49535V
NOVUNBP1-49535VEA 668 EUR
NOVUNBP1-49535V
Anti-RBBP6 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: RBBP6 (retinoblastoma binding protein 6)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR