Anti-DUX4 Mouse Monoclonal Antibody (DyLight® 680) [clone: P4H2]

Lieferant: Novus Biologicals
NBP1-49552FR
NOVUNBP1-49552FREA 650 EUR
NOVUNBP1-49552FR
Anti-DUX4 Mouse Monoclonal Antibody (DyLight® 680) [clone: P4H2]
Antikörper
Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality: monoclonal
Clone: P4H2
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR