Anti-KDM4A Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-49602B
NOVUNBP1-49602BEA 668 EUR
NOVUNBP1-49602B
Anti-KDM4A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: KDM4A (lysine (K)-specific demethylase 4A)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse