Anti-PINK1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-49678B
NOVUNBP1-49678BEA 723 EUR
NOVUNBP1-49678B
Anti-PINK1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse