Anti-PINK1 Rabbit Polyclonal Antibody (DyLight® 488)
Lieferant: Novus Biologicals
NBP1-49678G
NOVUNBP1-49678GEA 723 EUR
NOVUNBP1-49678G
Anti-PINK1 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: PINK1 (PTEN induced putative kinase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse