Anti-PTH1R Mouse Monoclonal Antibody

Lieferant: Novus Biologicals
NBP1-51640
NOVUNBP1-51640EA 612 EUR
NOVUNBP1-51640
Anti-PTH1R Mouse Monoclonal Antibody
Antikörper
Mouse Monoclonal Parathyroid Hormone Receptor 1 Antibody (4D2). Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: Parathyroid Hormone Receptor 1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR