Anti-MFN1 Rabbit Polyclonal Antibody (DyLight® 550)
Lieferant: Novus Biologicals
NBP1-51841R
NOVUNBP1-51841REA 723 EUR
NOVUNBP1-51841R
Anti-MFN1 Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Primary
Antigen: MFN1 (mitofusin 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat