Anti-ATF5 Goat Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-51908
NOVUNBP1-51908EA 624 EUR
NOVUNBP1-51908
Anti-ATF5 Goat Polyclonal Antibody
Antikörper
The ATF5 Antibody from Novus Biologicals is a goat polyclonal antibody to ATF5. This antibody reacts with human. The ATF5 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Peptide

Type: Primary
Antigen: ATF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: EVYKARSQRTRSC
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR