Anti-STK33 Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Novus Biologicals
NBP1-54156G
NOVUNBP1-54156GEA 668 EUR
NOVUNBP1-54156G
Anti-STK33 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: STK33 (serine/threonine kinase 33)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR