Anti-SLC15A4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-59636
NOVUNBP1-59636EA 639 EUR
NOVUNBP1-59636
Anti-SLC15A4 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal SLC15A4 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: SLC15A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR