Anti-F2R Mouse Monoclonal Antibody (Biotin) [clone: N2-11]

Lieferant: Novus Biologicals
NBP1-71770B
NOVUNBP1-71770BEA 722 EUR
NOVUNBP1-71770B
Anti-F2R Mouse Monoclonal Antibody (Biotin) [clone: N2-11]
Antikörper
Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: monoclonal
Clone: N2-11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR