Anti-MFN1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 11E9-1H12]

Lieferant: Novus Biologicals
NBP1-71775IR
NOVUNBP1-71775IREA 722 EUR
NOVUNBP1-71775IR
Anti-MFN1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 11E9-1H12]
Antikörper
Type: Primary
Antigen: MFN1 (mitofusin 1)
Clonality: monoclonal
Clone: 11E9-1H12
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR