Anti-IL17 Rat Monoclonal Antibody [clone: TC11-18H10]

Lieferant: Novus Biologicals
NBP1-72027
NOVUNBP1-72027EA 750 EUR
NOVUNBP1-72027
Anti-IL17 Rat Monoclonal Antibody [clone: TC11-18H10]
Antikörper
Rat Monoclonal IL17 Antibody. Tested Applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA. Tested Reactivity: Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: IL17
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR