Anti-CHD7 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-77393B
NOVUNBP1-77393BEA 722 EUR
NOVUNBP1-77393B
Anti-CHD7 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CHD7 (chromodomain helicase DNA binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse