Anti-CHD7 Rabbit Polyclonal Antibody (DyLight® 405)
Lieferant: Novus Biologicals
NBP1-77393V
NOVUNBP1-77393VEA 722 EUR
NOVUNBP1-77393V
Anti-CHD7 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: CHD7 (chromodomain helicase DNA binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse