Anti-HSPA13 Rabbit Polyclonal Antibody (DyLight® 550)
Lieferant: Novus Biologicals
NBP1-77456R
NOVUNBP1-77456REA 668 EUR
NOVUNBP1-77456R
Anti-HSPA13 Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human