Anti-TPH2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NBP1-77556B
NOVUNBP1-77556BEA 524 EUR
NOVUNBP1-77556B
Anti-TPH2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR