Anti-TPH2 Rabbit Polyclonal Antibody (DyLight® 488)
Lieferant: Novus Biologicals
NBP1-77556G
NOVUNBP1-77556GEA 524 EUR
NOVUNBP1-77556G
Anti-TPH2 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human