Anti-TPH2 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-77559B
NOVUNBP1-77559BEA 524 EUR
NOVUNBP1-77559B
Anti-TPH2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human