Anti-Hsp90B Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NBP1-77561F
NOVUNBP1-77561FEA 668 EUR
NOVUNBP1-77561F
Anti-Hsp90B Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: Hsp90B
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR