Anti-RBFOX3 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-77686B
NOVUNBP1-77686BEA 758 EUR
NOVUNBP1-77686B
Anti-RBFOX3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: RBFOX3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat