Anti-KDM2A Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NBP1-78305B
NOVUNBP1-78305BEA 668 EUR
NOVUNBP1-78305B
Anti-KDM2A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: KDM2A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR