Anti-GSAP Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NBP1-78400B
NOVUNBP1-78400BEA 524 EUR
NOVUNBP1-78400B
Anti-GSAP Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: GSAP (gamma-secretase activating protein)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human