Anti-HTR4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NBP1-78403B
NOVUNBP1-78403BEA 524 EUR
NOVUNBP1-78403B
Anti-HTR4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: HTR4 (5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR