Anti-MAS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Novus Biologicals
NBP1-78444H
NOVUNBP1-78444HEA 814 EUR
NOVUNBP1-78444H
Anti-MAS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: MAS1 (MAS1 oncogene)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR