Anti-PIEZO1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-78537-A NBP1-78537-B
NOVUNBP1-78537-AEA 735 EUR
NOVUNBP1-78537-A NOVUNBP1-78537-B
Anti-PIEZO1 Rabbit Polyclonal Antibody
Antikörper
The PIEZO1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PIEZO1. This antibody reacts with human, mouse. The PIEZO1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: PIEZO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR