Anti-LC3I Rabbit Polyclonal Antibody (DyLight® 550)

Lieferant: Novus Biologicals
NBP1-78964R
NOVUNBP1-78964REA 668 EUR
NOVUNBP1-78964R
Anti-LC3I Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Primary
Antigen: LC3I
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR