Anti-TET1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-78965 NBP1-78965SS
NOVUNBP1-78965EA 567 EUR
NOVUNBP1-78965 NOVUNBP1-78965SS
Anti-TET1 Rabbit Polyclonal Antibody
Antikörper
The TET1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TET1. This antibody reacts with mouse. The TET1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC / IF (Negative).

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: TET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR