Anti-TET1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Novus Biologicals
NBP1-78965H
NOVUNBP1-78965HEA 722 EUR
NOVUNBP1-78965H
Anti-TET1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: TET1 (tet methylcytosine dioxygenase 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR